Assay Details
Target Gene Details
Entrez Gene ID: | 84656 |
Gene Name: | glyoxylate reductase 1 homolog |
Gene Aliases: |
BM045, HIBDL, N-PAC, NP60 |
Location: |
Chr.16:4803203-4847388 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26756 | Chr.16:4772529 - 4855231 on Build GRCh38 | Gain+Loss | GLYR1 SEPT12 UBN1 SMIM22 ROGDI |
esv2422516 | Chr.16:4775920 - 5160979 on Build GRCh38 | Deletion | GLYR1 EEF2KMT SEPT12 UBN1 ALG1 SEC14L5 NAGPA-AS1 SMIM22 C16orf89 ROGDI PPL NAGPA |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | NMRAL1 LOC105371063 UBALD1 MGRN1 SMIM22 ROGDI ANKS3 PAM16 GLYR1 ADCY9 C16orf96 ALG1 CDIP1 NUDT16L1 C16orf89 VASN UBN1 TFAP4 GLIS2 DNAJA3 SRL MIR6769A EEF2KMT SEPT12 ZNF500 CORO7 SEC14L5 RBFOX1 NAGPA-AS1 LINC01569 PPL C16orf71 GLIS2-AS1 NAGPA HMOX2 CORO7-PAM16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |