Assay Details
Target Gene Details
Entrez Gene ID: | 1632 |
Gene Name: | enoyl-CoA delta isomerase 1 |
Gene Aliases: |
DCI |
Location: |
Chr.16:2239872-2251601 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ECI1 | NM_001178029.1 | NP_001171500.1 | ||
NM_001919.3 | NP_001910.2 | |||
AK291127.1 | ||||
BC000762.2 | AAH00762.1 | |||
BC002746.2 | AAH02746.1 | |||
BC009631.1 | AAH09631.1 | |||
BC019316.1 | AAH19316.1 | |||
BC020228.1 | AAH20228.1 | |||
CD675473.1 | ||||
L24774.1 | AAA35485.1 | |||
Z25820.1 | CAA81065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | FAM173A LOC105371047 AXIN1 LOC105371045 C1QTNF8 HBA2 CHTF18 NME3 ERVK13-1 HN1L TPSD1 MPG PIGQ DNASE1L2 IFT140 LOC107984876 HBA1 MSLN GNPTG POLR3K TSR3 RNPS1 CACNA1H CCDC154 MIR3176 LOC729652 TBC1D24 RHOT2 NTHL1 NME4 WDR90 KCTD5 HBZ SPSB3 CRAMP1 WFIKKN1 MIR1225 GNG13 MIR6511B1 PDPK1 RNF151 ZNF598 LMF1-AS1 MIR6768 MIR3178 RPL23AP5 HBQ1 SNORD60 RGS11 LOC105371184 MIR3180-5 FLJ42627 UNKL MIR662 GFER LINC00254 SNORA10 PRSS27 BRICD5 LOC652276 WDR24 RPL3L HBM RHBDF1 PRR35 RHBDL1 PDIA2 RAB40C CASKIN1 CLCN7 PRSS29P MIR6767 TELO2 E4F1 MRPS34 TMEM8A ATP6V0C LOC100287175 LOC105371038 LMF1 RPS2 STUB1 TSC2 ABCA3 HS3ST6 NARFL ARHGDIG MCRIP2 TRAF7 MIR3177 TMEM204 MRPL28 RAB26 SSTR5-AS1 WASIR2 MSRB1 LOC105371049 TPSB2 LOC106660606 LINC00235 ECI1 LOC100134368 PRR25 RAB11FIP3 FBXL16 PGP MIR940 LUC7L PTX4 SOX8 IGFALS HAGH MAPK8IP3 MIR5587 CCNF MIR6859-4 LOC105371046 EME2 PKD1 C16orf59 SNHG19 FAM234A CAPN15 METTL26 DECR2 HAGHL FAHD1 SLC9A3R2 AMDHD2 BAIAP3 METRN MLST8 NHLRC4 UBE2I NUBP2 JMJD8 NOXO1 SNORA78 MIR3677 ABCA17P NPW NPRL3 LOC107987233 CCDC78 NDUFB10 MIR4516 MEIOB SYNGR3 TPSG1 LOC101929280 DDX11L10 MIR4717 SNRNP25 TBL3 SSTR5 CEMP1 SNHG9 TPSAB1 RPUSD1 SNORA64 NTN3 C16orf91 |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | ZNF598 TSC2 PKD1 SNORD60 ABCA3 MIR3180-5 SNHG19 TRAF7 GFER DNASE1L2 SLC9A3R2 RAB26 MLST8 BRICD5 RNPS1 LOC105371049 LOC106660606 NOXO1 MIR3677 NTHL1 ECI1 NPW MIR4516 CASKIN1 SYNGR3 MIR1225 PGP MIR940 E4F1 MIR6511B1 MIR4717 TBL3 |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss | LOC106660606 MIR3677 ECI1 SNORD60 CASKIN1 SNHG19 TRAF7 PGP MIR940 E4F1 DNASE1L2 RAB26 MLST8 BRICD5 RNPS1 |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss | PGP MIR940 E4F1 LOC106660606 MIR3677 DNASE1L2 ECI1 CASKIN1 TRAF7 MLST8 BRICD5 RNPS1 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | MIR6768 MIR3178 TSC2 PKD1 SNORD60 ABCA3 MIR3180-5 C16orf59 SNHG19 TRAF7 DNASE1L2 AMDHD2 RAB26 MLST8 BRICD5 LOC652276 RNPS1 LOC105371049 LOC729652 TBC1D24 LOC106660606 MIR3677 ECI1 ABCA17P MIR4516 CASKIN1 MIR6767 MIR1225 PGP MIR940 E4F1 MIR6511B1 MIR4717 ATP6V0C CEMP1 PDPK1 NTN3 CCNF |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion | LOC106660606 MIR3677 ECI1 SNORD60 ABCA3 CASKIN1 SNHG19 TRAF7 PGP MIR940 E4F1 MIR4717 DNASE1L2 RAB26 MLST8 BRICD5 RNPS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |