Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | LOC105371038 LOC105371045 LOC101929280 METRN MRPL28 MAPK8IP3 UBE2I CAPN15 HAGHL PIGQ HBZ TMEM8A LOC107987233 MCRIP2 MIR6859-4 RHBDL1 ARHGDIG MIR6768 ERVK13-1 RHOT2 NPRL3 LOC105371184 CCDC154 NPW SLC9A3R2 MIR5587 LUC7L SSTR5 MIR6511B1 LOC652276 UNKL RAB40C LOC105371049 C1QTNF8 TBC1D24 WFIKKN1 FAM234A ABCA17P IFT140 NTHL1 MIR3176 TBL3 BAIAP3 NME3 WDR90 CHTF18 SNORA10 ABCA3 DNASE1L2 GFER LOC100134368 JMJD8 TMEM204 HS3ST6 RPL23AP5 RNPS1 PRR35 NOXO1 RAB26 IGFALS BRICD5 MIR3178 RAB11FIP3 LOC105371047 TSR3 PGP LINC00254 MIR3677 MIR3180-5 AMDHD2 HBA1 RGS11 HBM E4F1 RPL3L FLJ42627 POLR3K MSLN HBQ1 LOC729652 MIR940 TPSB2 KCTD5 C16orf91 MPG RNF151 FBXL16 GNG13 WASIR2 CRAMP1 TPSD1 MSRB1 RHBDF1 DDX11L10 MLST8 MRPS34 LOC100287175 TSC2 HBA2 NDUFB10 SNORD60 HAGH SNHG9 WDR24 NHLRC4 FAHD1 TRAF7 CLCN7 RPS2 LOC106660606 LMF1 LINC00235 CASKIN1 TELO2 MIR4516 PDIA2 DECR2 RPUSD1 FAM173A ZNF598 LMF1-AS1 PRSS27 PTX4 PRR25 CACNA1H SNHG19 ECI1 PDPK1 CEMP1 LOC107984876 CCDC78 MIR1225 MIR662 GNPTG PKD1 SNRNP25 SNORA64 NUBP2 NTN3 MIR3177 LOC105371046 SYNGR3 MIR4717 PRSS29P NME4 MIR6767 SSTR5-AS1 NARFL MEIOB STUB1 AXIN1 SPSB3 SNORA78 METTL26 ATP6V0C TPSAB1 C16orf59 SOX8 TPSG1 CCNF EME2 HN1L |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | PDIA2 DECR2 AXIN1 LOC100134368 RAB11FIP3 MRPL28 TMEM8A RGS11 RPL23AP5 NME4 ARHGDIG FAM234A |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | LOC105371038 RPUSD1 FAM173A MIR3176 LOC101929280 METRN PRR25 WDR90 FBXL16 CHTF18 CAPN15 HAGHL GNG13 PIGQ JMJD8 CCDC78 LOC107987233 MIR662 MCRIP2 PRR35 RHBDL1 LOC100287175 RHOT2 LOC105371184 RAB11FIP3 MIR5587 NARFL STUB1 WDR24 NHLRC4 METTL26 RAB40C MSLN WFIKKN1 LINC00235 |
nsv1700 | Chr.16:457843 - 476640 on Build GRCh38 | Insertion | RAB11FIP3 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | DECR2 AXIN1 LOC100134368 RAB11FIP3 MRPL28 TMEM8A RPL23AP5 NME4 CAPN15 LINC00235 |
esv1003529 | Chr.16:455611 - 459074 on Build GRCh38 | Insertion | RAB11FIP3 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | PDIA2 DECR2 AXIN1 LOC100134368 RAB11FIP3 MRPL28 TMEM8A RGS11 RPL23AP5 NME4 ARHGDIG FAM234A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |