Assay Details
Target Gene Details
Entrez Gene ID: | 254528 |
Gene Name: | meiosis specific with OB domains |
Gene Aliases: |
C16orf73, gs129 |
Location: |
Chr.16:1833983-1872178 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEIOB | NM_001163560.2 | NP_001157032.1 | ||
NM_152764.2 | NP_689977.2 | |||
BC029829.1 | AAH29829.1 | |||
CD300613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833119 | Chr.16:1834308 - 1911484 on Build GRCh38 | Loss | FAHD1 LINC00254 MEIOB HS3ST6 |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | FAHD1 HAGH MEIOB |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | FAHD1 HAGH MEIOB |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | NHLRC4 CACNA1H HBZ SNHG9 LOC105371047 MIR6511B1 PRSS27 MRPL28 SNORA10 MIR4516 RAB40C SLC9A3R2 LOC107987233 LOC101929280 RNF151 METRN MIR4717 CEMP1 SNRNP25 CLCN7 LUC7L TSR3 LOC105371038 HAGHL ARHGDIG LOC106660606 SYNGR3 FLJ42627 NUBP2 LOC105371046 MIR6859-4 MRPS34 MLST8 LINC00254 NTN3 MIR6768 MIR1225 E4F1 FAM234A RPL3L NME4 MIR3178 SNORA64 TPSB2 MSRB1 DDX11L10 NPW UBE2I TSC2 LOC729652 NTHL1 CCDC154 PGP TRAF7 IGFALS PKD1 CAPN15 HBA2 WFIKKN1 DECR2 RNPS1 RGS11 PRR25 FBXL16 PRSS29P TBC1D24 MIR3177 LMF1 HBA1 TBL3 NARFL HAGH PDIA2 RAB11FIP3 MIR3180-5 PDPK1 CASKIN1 PTX4 NME3 HS3ST6 C16orf59 CCNF CCDC78 ZNF598 TMEM8A SNHG19 MEIOB MIR940 LOC105371049 AMDHD2 WASIR2 ABCA17P SOX8 MIR3176 HN1L UNKL NPRL3 ATP6V0C FAHD1 TELO2 MIR662 SNORA78 IFT140 LOC100287175 MIR6767 METTL26 PRR35 LOC107984876 RPL23AP5 HBQ1 TPSD1 PIGQ WDR90 TPSAB1 AXIN1 MAPK8IP3 EME2 GNPTG FAM173A LOC652276 MIR3677 RPS2 POLR3K MPG BAIAP3 TMEM204 SSTR5-AS1 RPUSD1 HBM GNG13 RHBDL1 LOC105371045 ERVK13-1 RAB26 KCTD5 CHTF18 C16orf91 LOC100134368 TPSG1 MIR5587 GFER LOC105371184 JMJD8 RHOT2 MCRIP2 STUB1 LMF1-AS1 MSLN DNASE1L2 C1QTNF8 SPSB3 LINC00235 RHBDF1 SSTR5 NDUFB10 ABCA3 CRAMP1 BRICD5 NOXO1 SNORD60 ECI1 WDR24 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | MIR3177 HN1L TMEM204 HAGH LOC105371047 NUBP2 IGFALS LOC105371046 SPSB3 NME3 FAHD1 MRPS34 TELO2 MAPK8IP3 EME2 CRAMP1 IFT140 MEIOB |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | FAHD1 HAGH MEIOB NUBP2 IGFALS |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss | FAHD1 HAGH MEIOB |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | FAHD1 LINC00254 HAGH MEIOB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |