Assay Details
Target Gene Details
Entrez Gene ID: | 54620 |
Gene Name: | F-box and leucine rich repeat protein 19 |
Gene Aliases: |
CXXC11, Fbl19, JHDM1C |
Location: |
Chr.16:30923055-30948783 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBXL19 | NM_001099784.2 | NP_001093254.2 | ||
NM_001282351.1 | NP_001269280.1 | |||
AK098777.1 | BAC05410.1 | |||
AK127701.1 | ||||
AK302669.1 | ||||
BC059173.1 | AAH59173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv457483 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss | LOC101928762 MIR762HG MIR762 VKORC1 PRSS53 CTF1 FUS STX1B HSD3B7 FBXL19 BCL7C KAT8 ZNF646 PRSS36 FBXL19-AS1 SETD1A ORAI3 PRSS8 BCKDK STX4 ZNF668 |
dgv35n68 | Chr.16:30833730 - 31036730 on Build GRCh38 | Loss | MIR762HG MIR762 MIR4519 ORAI3 CTF1 STX4 STX1B HSD3B7 FBXL19 BCL7C FBXL19-AS1 SETD1A |
nsv953802 | Chr.16:30876680 - 30999279 on Build GRCh38 | Deletion | MIR762HG MIR762 ORAI3 CTF1 STX1B HSD3B7 FBXL19 BCL7C FBXL19-AS1 SETD1A |
nsv833189 | Chr.16:30808387 - 30949474 on Build GRCh38 | Loss | MIR762HG MIR762 MIR4519 ORAI3 CTF1 FBXL19 BCL7C FBXL19-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |