Assay Details
Target Gene Details
Entrez Gene ID: | 8140 |
Gene Name: | solute carrier family 7 member 5 |
Gene Aliases: |
4F2LC, CD98, D16S469E, E16, LAT1, MPE16, hLAT1 |
Location: |
Chr.16:87830022-87869499 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC7A5 | NM_003486.5 | NP_003477.4 | ||
XM_006721286.3 | XP_006721349.2 | |||
XM_017023735.1 | XP_016879224.1 | |||
XM_017023736.1 | XP_016879225.1 | |||
XM_017023737.1 | XP_016879226.1 | |||
AB017908.1 | BAA75746.1 | |||
AB018009.1 | BAA84648.1 | |||
AB018542.1 | BAA33851.1 | |||
AF077866.1 | AAC61479.1 | |||
AF104032.1 | AAD20464.1 | |||
BC039692.2 | AAH39692.1 | |||
BC042600.1 | AAH42600.1 | |||
M80244.1 | AAA35780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059786 | Chr.16:87811419 - 88019505 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv833322 | Chr.16:87715029 - 87884870 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv1160441 | Chr.16:87761248 - 87867260 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() |
esv3892935 | Chr.16:87808912 - 88010269 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv573557 | Chr.16:87808535 - 87854711 on Build GRCh38 | Loss |
![]() ![]() |
dgv3062n100 | Chr.16:87725235 - 87842257 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
dgv36n68 | Chr.16:87801736 - 87983550 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv1062946 | Chr.16:87635801 - 88019505 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv457616 | Chr.16:87705252 - 87914409 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)