Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | PDIA2 WDR24 SYNGR3 RGS11 RNPS1 PRR35 LMF1-AS1 LINC00254 MSLN IFT140 LOC729652 BAIAP3 FAHD1 RHBDF1 PRSS29P NHLRC4 CEMP1 SNRNP25 NME3 HN1L NARFL CLCN7 METTL26 LUC7L MIR3177 HS3ST6 MIR5587 LOC100134368 NTN3 AMDHD2 SOX8 SNORA64 TBL3 MCRIP2 CCDC78 UBE2I TMEM204 ECI1 HBM LOC106660606 FAM173A PDPK1 MRPS34 MIR6768 MAPK8IP3 CRAMP1 MIR6859-4 JMJD8 NTHL1 RAB11FIP3 PRSS27 ERVK13-1 LOC652276 TPSB2 TMEM8A HBZ LOC105371045 METRN RNF151 TSR3 ABCA3 AXIN1 HBA2 MIR662 LOC105371049 KCTD5 ZNF598 LOC105371046 NME4 DECR2 HAGHL RAB26 WDR90 PKD1 CHTF18 MIR940 TRAF7 SNORA10 LMF1 GNG13 GNPTG PRR25 RHBDL1 MIR6511B1 HBA1 TPSAB1 TPSG1 MRPL28 NPW DNASE1L2 ATP6V0C MIR1225 RPUSD1 RPL23AP5 CCDC154 MIR3178 TPSD1 TELO2 WASIR2 LINC00235 RHOT2 SNHG9 C16orf91 TBC1D24 SSTR5 EME2 CCNF FLJ42627 MIR3180-5 PTX4 STUB1 LOC105371047 LOC105371038 RPS2 LOC107987233 SNORD60 RAB40C CAPN15 UNKL SNHG19 CASKIN1 SSTR5-AS1 C16orf59 MIR3176 ARHGDIG DDX11L10 MIR4516 MIR3677 SNORA78 SPSB3 CACNA1H C1QTNF8 RPL3L IGFALS MPG LOC105371184 FBXL16 MSRB1 HAGH GFER E4F1 FAM234A NPRL3 LOC101929280 BRICD5 LOC100287175 MLST8 TSC2 MIR4717 SLC9A3R2 MEIOB HBQ1 NUBP2 MIR6767 NOXO1 ABCA17P PGP POLR3K NDUFB10 PIGQ WFIKKN1 LOC107984876 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | FAM234A MRPL28 LOC100134368 PDIA2 TMEM8A NME4 DECR2 RGS11 RPL23AP5 RAB11FIP3 AXIN1 ARHGDIG |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | LINC00235 MRPL28 LOC100134368 TMEM8A CAPN15 NME4 DECR2 RPL23AP5 RAB11FIP3 AXIN1 |
dgv4809n54 | Chr.16:322597 - 451362 on Build GRCh38 | Loss | MRPL28 LOC100134368 TMEM8A NME4 DECR2 RPL23AP5 RAB11FIP3 AXIN1 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | FAM234A MRPL28 LOC100134368 PDIA2 TMEM8A NME4 DECR2 RGS11 RPL23AP5 RAB11FIP3 AXIN1 ARHGDIG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |