Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | CCNF TRAF7 HBA1 IFT140 LOC105371047 METRN LOC105371049 PRR35 NPRL3 DDX11L10 NDUFB10 WASIR2 CHTF18 MIR3178 C16orf91 C1QTNF8 RAB11FIP3 UBE2I NARFL LOC105371184 LINC00235 SPSB3 PDPK1 RPUSD1 GFER ABCA17P TMEM8A NUBP2 HBM WDR24 CRAMP1 RHBDL1 MPG DECR2 LOC100287175 RNPS1 CLCN7 SNRNP25 STUB1 CCDC154 WDR90 SNORD60 HAGH MIR6511B1 DNASE1L2 PRR25 PRSS29P PGP BRICD5 NPW HS3ST6 NME4 HBQ1 IGFALS RHBDF1 MIR4516 PTX4 TMEM204 MIR3177 RHOT2 FLJ42627 RGS11 MIR6767 PDIA2 SSTR5 LMF1 CEMP1 MSRB1 LOC107987233 LOC105371045 WFIKKN1 TSR3 ATP6V0C TPSB2 MIR662 PIGQ TBL3 KCTD5 E4F1 METTL26 FBXL16 HN1L MLST8 FAHD1 CACNA1H LOC105371038 MIR5587 RAB40C MIR6859-4 MEIOB TPSAB1 RPS2 UNKL HBZ SSTR5-AS1 MIR6768 PKD1 NOXO1 PRSS27 RPL23AP5 LOC100134368 TPSD1 RNF151 FAM173A CAPN15 CASKIN1 GNG13 LOC107984876 MIR1225 SOX8 SLC9A3R2 AMDHD2 MAPK8IP3 LOC729652 RPL3L MCRIP2 MSLN NHLRC4 ERVK13-1 LOC652276 FAM234A MRPS34 LOC105371046 MIR4717 LINC00254 ZNF598 ARHGDIG LOC101929280 POLR3K MIR3176 C16orf59 ECI1 ABCA3 RAB26 CCDC78 JMJD8 HBA2 SNHG9 SNORA78 MIR3677 LOC106660606 GNPTG TBC1D24 SNHG19 LMF1-AS1 AXIN1 MIR940 SNORA10 MRPL28 SNORA64 BAIAP3 EME2 HAGHL TSC2 NME3 SYNGR3 NTN3 TPSG1 NTHL1 MIR3180-5 TELO2 LUC7L |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | RPL23AP5 AXIN1 LOC100134368 FAM234A NME4 RGS11 TMEM8A DECR2 MRPL28 PDIA2 RAB11FIP3 ARHGDIG |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | RPL23AP5 AXIN1 LOC100134368 NME4 CAPN15 TMEM8A DECR2 MRPL28 RAB11FIP3 LINC00235 |
dgv4809n54 | Chr.16:322597 - 451362 on Build GRCh38 | Loss | RPL23AP5 AXIN1 LOC100134368 NME4 TMEM8A DECR2 MRPL28 RAB11FIP3 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | RPL23AP5 AXIN1 LOC100134368 FAM234A NME4 RGS11 TMEM8A DECR2 MRPL28 PDIA2 RAB11FIP3 ARHGDIG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |