Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521610 | Chr.16:430096 - 445538 on Build GRCh38 | Gain | RAB11FIP3 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MAPK8IP3 HBQ1 SNHG9 HAGHL ARHGDIG BRICD5 MIR4516 ABCA3 ATP6V0C SNHG19 UNKL PIGQ LOC729652 FLJ42627 LINC00235 HBA1 LMF1-AS1 LOC107987233 NTHL1 MIR1225 SYNGR3 LINC00254 LOC105371038 NUBP2 TMEM8A CCDC154 MRPL28 CASKIN1 NTN3 LOC100134368 RNPS1 LOC107984876 RPUSD1 PDPK1 METRN CCNF NPRL3 SNORA10 SOX8 LMF1 TBC1D24 RHOT2 FAM234A NHLRC4 SPSB3 SLC9A3R2 ECI1 HBA2 E4F1 DNASE1L2 METTL26 TPSAB1 IGFALS TMEM204 TPSB2 MIR662 DECR2 MIR3177 AXIN1 MSLN ERVK13-1 PRSS27 PDIA2 RPL3L WDR24 HS3ST6 RHBDF1 TELO2 PTX4 GFER TPSD1 C16orf91 WDR90 PRR35 PGP SNRNP25 NDUFB10 HAGH MLST8 RNF151 MIR4717 CCDC78 RPL23AP5 WFIKKN1 MRPS34 RPS2 EME2 TSR3 RAB11FIP3 ZNF598 RAB40C GNG13 SSTR5-AS1 FAM173A HN1L PRSS29P JMJD8 PRR25 NARFL CRAMP1 TPSG1 SNORD60 MIR6859-4 MIR6768 MPG CEMP1 LOC105371049 LOC105371046 TSC2 MIR6511B1 HBM FBXL16 SNORA64 MIR6767 AMDHD2 CLCN7 MIR5587 NOXO1 C16orf59 LOC105371184 MIR3180-5 MIR3178 NPW RGS11 PKD1 ABCA17P TBL3 FAHD1 STUB1 LOC100287175 CACNA1H LOC652276 C1QTNF8 NME4 GNPTG MSRB1 BAIAP3 IFT140 CAPN15 DDX11L10 MIR940 RHBDL1 HBZ LOC105371045 TRAF7 WASIR2 POLR3K NME3 CHTF18 MCRIP2 MEIOB LOC105371047 MIR3677 SNORA78 LOC106660606 LOC101929280 LUC7L MIR3176 KCTD5 SSTR5 UBE2I RAB26 |
dgv4810n54 | Chr.16:380011 - 445662 on Build GRCh38 | Loss | DECR2 RPL23AP5 TMEM8A LOC100134368 RAB11FIP3 NME4 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | DECR2 RGS11 RPL23AP5 TMEM8A AXIN1 MRPL28 LOC100134368 PDIA2 RAB11FIP3 ARHGDIG FAM234A NME4 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | LINC00235 DECR2 CAPN15 RPL23AP5 TMEM8A AXIN1 MRPL28 LOC100134368 RAB11FIP3 NME4 |
nsv457303 | Chr.16:383623 - 445662 on Build GRCh38 | Loss | DECR2 RPL23AP5 LOC100134368 RAB11FIP3 NME4 |
dgv4809n54 | Chr.16:322597 - 451362 on Build GRCh38 | Loss | DECR2 RPL23AP5 TMEM8A AXIN1 MRPL28 LOC100134368 RAB11FIP3 NME4 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | DECR2 RGS11 RPL23AP5 TMEM8A AXIN1 MRPL28 LOC100134368 PDIA2 RAB11FIP3 ARHGDIG FAM234A NME4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |