Assay Details
Target Gene Details
Entrez Gene ID: | 54715 |
Gene Name: | RNA binding protein, fox-1 homolog 1 |
Gene Aliases: |
2BP1, A2BP1, FOX-1, FOX1, HRNBP1 |
Location: |
Chr.16:5239749-7713343 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBFOX1 | XM_017023319.1 | XP_016878808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035156 | Chr.16:5091032 - 5996934 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1045304 | Chr.16:5205761 - 5326875 on Build GRCh38 | Loss |
![]() |
dgv2698n100 | Chr.16:5304641 - 5327717 on Build GRCh38 | Loss |
![]() |
nsv1036241 | Chr.16:5271882 - 5387609 on Build GRCh38 | Loss |
![]() |
esv34300 | Chr.16:5005938 - 5322534 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3892764 | Chr.16:5316436 - 5404485 on Build GRCh38 | Loss |
![]() |
esv2760069 | Chr.16:4922849 - 5374828 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428318 | Chr.16:5011594 - 5374828 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2762021 | Chr.16:5293419 - 5377238 on Build GRCh38 | Loss |
![]() |
nsv1037303 | Chr.16:5287645 - 5327595 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)