Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | PGP IGFALS PRR25 MSRB1 SYNGR3 NUBP2 MIR3180-5 HBQ1 METRN SPSB3 TRAF7 MIR940 C1QTNF8 NARFL GFER TBC1D24 EME2 LOC107984876 MIR4717 MRPS34 RAB11FIP3 FAHD1 GNPTG NDUFB10 IFT140 LMF1 LOC105371045 HAGHL C16orf59 RNPS1 RPS2 MIR4516 ZNF598 WDR24 MIR3677 CEMP1 LOC105371184 MIR3177 MIR3178 PRSS29P CCNF ABCA3 SNHG9 KCTD5 MPG LMF1-AS1 CASKIN1 MRPL28 HBM NME3 METTL26 PDIA2 WASIR2 PDPK1 MIR6768 NTHL1 HBA2 SNORA78 FAM234A TSC2 SSTR5 LOC105371047 MIR3176 PTX4 LINC00235 NPRL3 MIR6511B1 RGS11 TPSB2 PRSS27 FAM173A POLR3K LOC105371046 JMJD8 MIR662 LOC105371038 CACNA1H SSTR5-AS1 ATP6V0C LOC105371049 LOC101929280 AMDHD2 RPL23AP5 UNKL RPUSD1 RHBDL1 MAPK8IP3 FLJ42627 CCDC78 STUB1 MIR5587 MSLN BAIAP3 LOC100134368 RHBDF1 LUC7L LOC652276 FBXL16 SNRNP25 SOX8 SLC9A3R2 RAB40C MIR1225 CHTF18 NPW WFIKKN1 TPSAB1 ARHGDIG CCDC154 RNF151 CRAMP1 SNORA10 SNORA64 CAPN15 MIR6859-4 HN1L TPSD1 LOC106660606 HS3ST6 TMEM204 DECR2 TBL3 HAGH NME4 PRR35 MLST8 TSR3 GNG13 ERVK13-1 HBZ NOXO1 RPL3L TELO2 PKD1 LINC00254 WDR90 ABCA17P AXIN1 MIR6767 MEIOB ECI1 DNASE1L2 BRICD5 HBA1 DDX11L10 C16orf91 UBE2I RHOT2 NTN3 LOC729652 SNORD60 PIGQ TMEM8A RAB26 LOC107987233 LOC100287175 E4F1 CLCN7 NHLRC4 TPSG1 MCRIP2 SNHG19 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | RPL23AP5 ARHGDIG FAM234A RAB11FIP3 PDIA2 DECR2 NME4 LOC100134368 RGS11 AXIN1 TMEM8A MRPL28 |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | RPUSD1 PRR25 RHBDL1 CCDC78 STUB1 MIR5587 PRR35 MSLN GNG13 METRN METTL26 NARFL FBXL16 WDR90 RAB40C RAB11FIP3 MIR3176 CHTF18 LINC00235 RHOT2 WFIKKN1 PIGQ FAM173A LOC107987233 LOC100287175 HAGHL JMJD8 MIR662 LOC105371038 WDR24 CAPN15 NHLRC4 LOC105371184 MCRIP2 LOC101929280 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | RPL23AP5 RAB11FIP3 DECR2 LINC00235 NME4 CAPN15 LOC100134368 AXIN1 TMEM8A MRPL28 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | RPL23AP5 ARHGDIG FAM234A RAB11FIP3 PDIA2 DECR2 NME4 LOC100134368 RGS11 AXIN1 TMEM8A MRPL28 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |