Assay Details
Target Gene Details
Entrez Gene ID: | 51451 |
Gene Name: | leucine carboxyl methyltransferase 1 |
Gene Aliases: |
CGI-68, LCMT, PPMT1 |
Location: |
Chr.16:25111709-25178231 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LCMT1 | NM_001032391.1 | NP_001027563.1 | ||
NM_016309.2 | NP_057393.2 | |||
XM_005255354.3 | XP_005255411.1 | |||
XM_011545862.2 | XP_011544164.1 | |||
XM_011545864.1 | XP_011544166.1 | |||
AF037601.1 | AAF18267.1 | |||
AF151826.1 | AAD34063.1 | |||
AK025884.1 | BAB15270.1 | |||
AK223364.1 | BAD97084.1 | |||
AK225271.1 | ||||
AK291885.1 | ||||
AK314409.1 | ||||
AL137283.1 | CAB70677.1 | |||
AL560574.3 | ||||
BC001214.1 | AAH01214.1 | |||
BC014217.2 | AAH14217.1 | |||
BI459868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758638 | Chr.16:24932074 - 25175657 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)