Assay Details
Target Gene Details
Entrez Gene ID: | 1632 |
Gene Name: | enoyl-CoA delta isomerase 1 |
Gene Aliases: |
DCI |
Location: |
Chr.16:2239872-2251601 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ECI1 | NM_001178029.1 | NP_001171500.1 | ||
NM_001919.3 | NP_001910.2 | |||
AK291127.1 | ||||
BC000762.2 | AAH00762.1 | |||
BC002746.2 | AAH02746.1 | |||
BC009631.1 | AAH09631.1 | |||
BC019316.1 | AAH19316.1 | |||
BC020228.1 | AAH20228.1 | |||
CD675473.1 | ||||
L24774.1 | AAA35485.1 | |||
Z25820.1 | CAA81065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | PGP TSC2 SNORD60 SPSB3 RHBDF1 FAM234A MPG LOC105371045 RGS11 HBM RNF151 MIR6768 PKD1 SSTR5-AS1 MAPK8IP3 ERVK13-1 MIR3177 RPL23AP5 FAHD1 FBXL16 DDX11L10 FAM173A GFER C16orf59 PIGQ MIR3677 LINC00235 CACNA1H TSR3 UBE2I CCDC78 HN1L CRAMP1 NPRL3 NME3 PRR25 MIR940 NHLRC4 BAIAP3 IFT140 LOC729652 DNASE1L2 C16orf91 MIR3176 KCTD5 NTN3 MRPS34 SNORA64 LOC107987233 HBQ1 WFIKKN1 CHTF18 ECI1 TBC1D24 GNPTG SNORA10 MIR3180-5 CCDC154 MIR1225 RPS2 HAGH MSLN SNORA78 CASKIN1 MIR4516 NME4 MIR6511B1 HS3ST6 LOC106660606 MSRB1 NDUFB10 ATP6V0C MCRIP2 PTX4 TMEM204 TPSD1 MIR3178 NOXO1 HBA1 LOC105371046 STUB1 TBL3 ABCA17P TELO2 ABCA3 MIR5587 IGFALS AMDHD2 POLR3K SNHG19 MIR6767 MIR4717 DECR2 WDR90 WASIR2 FLJ42627 LOC105371038 RNPS1 GNG13 WDR24 EME2 C1QTNF8 MIR6859-4 HBZ TPSG1 MEIOB NTHL1 ZNF598 CCNF E4F1 NUBP2 AXIN1 RAB40C TMEM8A METRN PRSS29P METTL26 NPW LOC105371049 TPSB2 CEMP1 MRPL28 LOC100134368 NARFL LINC00254 HAGHL TRAF7 LUC7L LOC101929280 RAB11FIP3 RPL3L CAPN15 LMF1-AS1 LOC105371047 LOC652276 PRR35 RHOT2 LOC100287175 JMJD8 RPUSD1 LOC105371184 PRSS27 TPSAB1 PDPK1 SNRNP25 SSTR5 LOC107984876 SLC9A3R2 HBA2 MLST8 CLCN7 MIR662 LMF1 ARHGDIG SNHG9 RAB26 UNKL BRICD5 SOX8 RHBDL1 SYNGR3 PDIA2 |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | PGP TSC2 SNORD60 RNPS1 TRAF7 ECI1 PKD1 NTHL1 ZNF598 MIR3180-5 MIR1225 CASKIN1 MIR4516 MIR6511B1 E4F1 LOC106660606 SLC9A3R2 GFER NOXO1 MLST8 MIR3677 TBL3 ABCA3 SNHG19 MIR940 DNASE1L2 NPW RAB26 LOC105371049 MIR4717 BRICD5 SYNGR3 |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss | PGP LOC106660606 SNORD60 RNPS1 TRAF7 MLST8 ECI1 MIR3677 SNHG19 MIR940 DNASE1L2 RAB26 CASKIN1 BRICD5 E4F1 |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss | PGP LOC106660606 RNPS1 MIR940 TRAF7 DNASE1L2 MLST8 ECI1 MIR3677 CASKIN1 BRICD5 E4F1 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | PGP TSC2 SNORD60 RNPS1 TRAF7 ECI1 TBC1D24 MIR6768 LOC652276 PKD1 MIR3180-5 MIR1225 CCNF CASKIN1 MIR4516 PDPK1 MIR6511B1 E4F1 LOC106660606 ATP6V0C MIR3178 C16orf59 MLST8 MIR3677 ABCA17P ABCA3 AMDHD2 SNHG19 MIR940 LOC729652 DNASE1L2 RAB26 LOC105371049 MIR6767 MIR4717 CEMP1 NTN3 BRICD5 |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion | PGP LOC106660606 SNORD60 RNPS1 TRAF7 MLST8 ECI1 MIR3677 ABCA3 SNHG19 MIR940 DNASE1L2 RAB26 MIR4717 CASKIN1 BRICD5 E4F1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |