Assay Details
Target Gene Details
Entrez Gene ID: | 4016 |
Gene Name: | lysyl oxidase like 1 |
Gene Aliases: |
LOL, LOXL |
Location: |
Chr.15:73926451-73952141 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOXL1 | NM_005576.3 | NP_005567.2 | ||
XM_011521555.1 | XP_011519857.1 | |||
AK303982.1 | ||||
AK314222.1 | ||||
BC015090.2 | AAH15090.1 | |||
BC068542.1 | AAH68542.1 | |||
L21186.1 | AAA50162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1604 | Chr.15:73903279 - 73936860 on Build GRCh38 | Insertion | LOXL1-AS1 TBC1D21 LOXL1 |
nsv1603 | Chr.15:73898881 - 73943392 on Build GRCh38 | Deletion | LOXL1-AS1 TBC1D21 LOXL1 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | LOC732265 GOLGA6A PML LOXL1-AS1 LOC283731 STOML1 ISLR2 LOXL1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | C15orf59-AS1 CYP1A2 CSK C15orf39 REC114 NEO1 CLK3 UBL7 HIGD2B ADPGK HCN4 LOC105376731 NPTN PML STRA6 MPI LMAN1L SEMA7A CYP1A1 CYP11A1 GOLGA6C GOLGA6A SCAMP2 LOXL1-AS1 FAM219B RPP25 LOC102723750 UBL7-AS1 LOC729739 CD276 SCAMP5 ARID3B CPLX3 MIR4513 CCDC33 BBS4 C15orf59 COX5A MIR6882 TBC1D21 STOML1 ULK3 NPTN-IT1 LOC283731 ISLR LOC101929333 LOXL1 PPCDC LOC732265 ADPGK-AS1 EDC3 MIR6881 LOC107984728 ISLR2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |