Assay Details
Target Gene Details
Entrez Gene ID: | 2923 |
Gene Name: | protein disulfide isomerase family A member 3 |
Gene Aliases: |
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC |
Location: |
Chr.15:43746392-43772606 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDIA3 | NM_005313.4 | NP_005304.3 | ||
AK075455.1 | ||||
AK075481.1 | ||||
AK293249.1 | ||||
BC014433.1 | AAH14433.1 | |||
BC036000.1 | ||||
BC071878.1 | AAH71878.1 | |||
BG826781.1 | ||||
D16234.1 | BAA03759.1 | |||
D83485.1 | BAA11928.1 | |||
EU668338.1 | ||||
FJ224330.1 | ||||
U42068.1 | AAC50331.1 | |||
Z49835.1 | CAA89996.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2760028 | Chr.15:43530960 - 43856443 on Build GRCh38 | Gain+Loss | PPIP5K1 STRCP1 CATSPER2P1 SERF2-C15ORF63 HYPK SERF2 STRC CKMT1A CATSPER2 MFAP1 WDR76 PDIA3 MAP1A CKMT1B MIR1282 ELL3 SERINC4 |
esv33661 | Chr.15:43724321 - 43772222 on Build GRCh38 | Loss | PDIA3 CATSPER2P1 |
esv3569520 | Chr.15:43596064 - 43762536 on Build GRCh38 | Gain | PDIA3 STRCP1 CKMT1B CATSPER2P1 STRC CKMT1A CATSPER2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |