Assay Details
Target Gene Details
Entrez Gene ID: | 84465 |
Gene Name: | multiple EGF like domains 11 |
Gene Aliases: |
- |
Location: |
Chr.15:65895296-66253737 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEGF11 | NM_032445.2 | NP_115821.2 | ||
XM_017022670.1 | XP_016878159.1 | |||
XM_017022671.1 | XP_016878160.1 | |||
XM_017022672.1 | XP_016878161.1 | |||
XM_017022674.1 | XP_016878163.1 | |||
XM_017022675.1 | XP_016878164.1 | |||
AB058677.1 | ||||
AB300051.1 | ||||
AL834326.1 | CAD38994.1 | |||
AY358226.1 | AAQ88593.1 | |||
BC117419.1 | ||||
BC126313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833044 | Chr.15:65766302 - 65969046 on Build GRCh38 | Loss | DENND4A MEGF11 LOC646358 RAB11A |
nsv471684 | Chr.15:65890789 - 66030069 on Build GRCh38 | Gain | MEGF11 RAB11A |
esv2760035 | Chr.15:65888447 - 66058047 on Build GRCh38 | Loss | MEGF11 MIR4311 RAB11A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |