Assay Details
Target Gene Details
Entrez Gene ID: | 57538 |
Gene Name: | alpha kinase 3 |
Gene Aliases: |
MAK, MIDORI |
Location: |
Chr.15:84816631-84873482 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALPK3 | NM_020778.4 | NP_065829.3 | ||
XM_017022444.1 | XP_016877933.1 | |||
XM_017022445.1 | XP_016877934.1 | |||
AY044449.1 | AAK95951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2760049 | Chr.15:84719698 - 84873299 on Build GRCh38 | Loss | ZNF592 ALPK3 |
esv2672879 | Chr.15:84183834 - 85188520 on Build GRCh38 | Deletion | ZSCAN2 LOC440300 UBE2Q2P1 GOLGA2P7 UBE2Q2L GOLGA6L5P GOLGA6L4 LOC105376722 LOC102724135 SCAND2P SEC11A LOC103171574 ZNF592 CSPG4P5 WDR73 ALPK3 SLC28A1 PDE8A NMB LINC00933 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |