Assay Details
Target Gene Details
Entrez Gene ID: | 4016 |
Gene Name: | lysyl oxidase like 1 |
Gene Aliases: |
LOL, LOXL |
Location: |
Chr.15:73926451-73952141 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOXL1 | NM_005576.3 | NP_005567.2 | ||
XM_011521555.1 | XP_011519857.1 | |||
AK303982.1 | ||||
AK314222.1 | ||||
BC015090.2 | AAH15090.1 | |||
BC068542.1 | AAH68542.1 | |||
L21186.1 | AAA50162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1604 | Chr.15:73903279 - 73936860 on Build GRCh38 | Insertion | LOXL1-AS1 TBC1D21 LOXL1 |
nsv1603 | Chr.15:73898881 - 73943392 on Build GRCh38 | Deletion | LOXL1-AS1 TBC1D21 LOXL1 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | LOXL1-AS1 LOXL1 GOLGA6A LOC283731 LOC732265 STOML1 ISLR2 PML |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | ISLR ADPGK GOLGA6C REC114 MIR6882 STOML1 LOC107984728 ARID3B LMAN1L C15orf59 GOLGA6A BBS4 FAM219B LOC102723750 PPCDC CYP11A1 ADPGK-AS1 COX5A CD276 HIGD2B LOXL1-AS1 C15orf39 LOC105376731 CYP1A2 LOC729739 ULK3 ISLR2 HCN4 C15orf59-AS1 LOC101929333 UBL7 MPI CLK3 UBL7-AS1 CPLX3 STRA6 MIR4513 MIR6881 CCDC33 TBC1D21 NPTN-IT1 LOC283731 SEMA7A LOC732265 NPTN PML NEO1 CYP1A1 LOXL1 SCAMP5 EDC3 CSK RPP25 SCAMP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |