Assay Details
Target Gene Details
Entrez Gene ID: | 4016 |
Gene Name: | lysyl oxidase like 1 |
Gene Aliases: |
LOL, LOXL |
Location: |
Chr.15:73926451-73952141 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOXL1 | NM_005576.3 | NP_005567.2 | ||
XM_011521555.1 | XP_011519857.1 | |||
AK303982.1 | ||||
AK314222.1 | ||||
BC015090.2 | AAH15090.1 | |||
BC068542.1 | AAH68542.1 | |||
L21186.1 | AAA50162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1604 | Chr.15:73903279 - 73936860 on Build GRCh38 | Insertion | LOXL1 LOXL1-AS1 TBC1D21 |
nsv1603 | Chr.15:73898881 - 73943392 on Build GRCh38 | Deletion | LOXL1 LOXL1-AS1 TBC1D21 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | LOC283731 STOML1 LOXL1 LOXL1-AS1 ISLR2 GOLGA6A LOC732265 PML |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | ADPGK UBL7 LOC729739 C15orf59-AS1 GOLGA6A CYP1A1 ULK3 FAM219B MIR6881 CPLX3 SCAMP5 NPTN-IT1 LOC283731 C15orf59 NEO1 ADPGK-AS1 REC114 GOLGA6C BBS4 EDC3 SEMA7A MPI LOC102723750 STOML1 PPCDC STRA6 RPP25 PML LOC107984728 MIR6882 CLK3 C15orf39 NPTN CYP1A2 ARID3B ISLR LOXL1 MIR4513 CYP11A1 CD276 SCAMP2 HIGD2B CCDC33 ISLR2 CSK LMAN1L COX5A TBC1D21 UBL7-AS1 LOC101929333 LOXL1-AS1 LOC105376731 LOC732265 HCN4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |