Assay Details
Target Gene Details
Entrez Gene ID: | 57519 |
Gene Name: | StAR related lipid transfer domain containing 9 |
Gene Aliases: |
KIF16A |
Location: |
Chr.15:42575646-42720998 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD9 | NM_020759.2 | NP_065810.2 | ||
XM_011521831.2 | XP_011520133.1 | |||
XM_011521832.2 | XP_011520134.1 | |||
XM_011521833.2 | XP_011520135.1 | |||
XM_011521834.2 | XP_011520136.1 | |||
XM_011521835.2 | XP_011520137.1 | |||
XM_011521837.2 | XP_011520139.1 | |||
XM_017022439.1 | XP_016877928.1 | |||
XM_017022440.1 | XP_016877929.1 | |||
CR627426.1 | CAH10513.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528725 | Chr.15:42463357 - 42812208 on Build GRCh38 | Gain | STARD9 CDAN1 ZNF106 LRRC57 SNAP23 TTBK2 HAUS2 |
esv3892679 | Chr.15:42568393 - 42596849 on Build GRCh38 | Loss | STARD9 HAUS2 |
esv2749624 | Chr.15:42569613 - 42595203 on Build GRCh38 | Deletion | STARD9 HAUS2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |