Assay Details
Target Gene Details
Entrez Gene ID: | 58489 |
Gene Name: | abhydrolase domain containing 17C |
Gene Aliases: |
FAM108C1 |
Location: |
Chr.15:80695311-80755621 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD17C | NM_021214.1 | NP_067037.1 | ||
AK092758.1 | ||||
AK298606.1 | ||||
AL390079.1 | CAB98203.1 | |||
BC059401.1 | ||||
BC115003.1 | AAI15004.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)