Assay Details
Target Gene Details
Entrez Gene ID: | 117153 |
Gene Name: | melanoma inhibitory activity 2 |
Gene Aliases: |
- |
Location: |
Chr.14:39233921-39253371 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MIA2 | NM_054024.3 | NP_473365.3 | ||
AF390175.2 | AAL26990.2 | |||
AK314069.1 | ||||
BC035981.1 | AAH35981.1 | |||
BC130537.1 | ||||
BC130563.1 | ||||
BX495701.1 | ||||
BX648228.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1053133 | Chr.14:39188331 - 39301525 on Build GRCh38 | Gain | CTAGE5 MIA2 LOC100288846 |
nsv84871 | Chr.14:39229196 - 39238793 on Build GRCh38 | Deletion | MIA2 |
esv3584293 | Chr.14:39191534 - 39339749 on Build GRCh38 | Gain | CTAGE5 MIA2 LOC100288846 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |