Assay Details
Target Gene Details
Entrez Gene ID: | 4140 |
Gene Name: | microtubule affinity regulating kinase 3 |
Gene Aliases: |
CTAK1, KP78, PAR1A, Par-1a |
Location: |
Chr.14:103385364-103503831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952258 | Chr.14:103386664 - 103405163 on Build GRCh38 | Duplication | MARK3 |
nsv9168 | Chr.14:103065392 - 103446969 on Build GRCh38 | Loss | LINC00677 MARK3 EIF5 LINC00605 LOC105378183 LOC105370687 EXOC3L4 SNORA28 TNFAIP2 LOC107984640 |
esv3635592 | Chr.14:103373908 - 103388946 on Build GRCh38 | Gain | MARK3 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | AKT1 TDRD9 BTBD6 PPP1R13B MIR6765 C14orf180 ZBTB42 C14orf79 LOC105370691 CKB APOPT1 PACS2 MIR4710 MIR203A LINC00638 TMEM179 ASPG LOC105370687 LOC101929634 SNORA28 LINC00637 JAG2 SIVA1 BAG5 INF2 LOC102723354 MARK3 EIF5 MIR203B LOC105370708 C14orf2 CDCA4 LOC107984648 CEP170B RD3L LOC105378183 ZFYVE21 GPR132 PLD4 AHNAK2 KIF26A BRF1 LOC388022 NUDT14 XRCC3 TRMT61A ADSSL1 KLC1 |
dgv422e214 | Chr.14:103355152 - 103437655 on Build GRCh38 | Gain | MARK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |