Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | CDCA4 C14orf79 GPR132 AHNAK2 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | CDCA4 LOC102723354 C14orf79 JAG2 GPR132 AHNAK2 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | CDCA4 C14orf79 PLD4 AHNAK2 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | C14orf79 PLD4 AHNAK2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | CDCA4 BTBD6 LOC107984648 C14orf2 MIR6765 GPR132 LOC105370691 ADSSL1 APOPT1 SIVA1 MIR4710 TMEM179 NUDT14 MIR203B C14orf180 ZFYVE21 C14orf79 TDRD9 ASPG LOC105370708 LOC101929634 PLD4 LOC105378183 ZBTB42 MIR203A MARK3 LOC102723354 KLC1 CEP170B JAG2 XRCC3 TRMT61A KIF26A AHNAK2 PPP1R13B AKT1 INF2 RD3L LINC00637 PACS2 SNORA28 CKB BRF1 BAG5 EIF5 LOC105370687 LINC00638 LOC388022 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | CDCA4 BTBD6 LOC102723354 MIR6765 C14orf79 JAG2 GPR132 BRF1 AHNAK2 NUDT14 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | CDCA4 LOC102723354 MIR6765 C14orf79 JAG2 GPR132 BRF1 AHNAK2 NUDT14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |