Assay Details
Target Gene Details
Entrez Gene ID: | 253970 |
Gene Name: | surfactant associated 3 |
Gene Aliases: |
NANCI, SFTPH, SP-H |
Location: |
Chr.14:36473288-36513785 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SFTA3 | NM_001101341.1 | NP_001094811.1 | ||
AY102071.1 | ||||
KF856952.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)