Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 | ||
XM_005267299.1 | XP_005267356.1 | |||
AK094143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | C14orf79 CDCA4 GPR132 AHNAK2 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | C14orf79 CDCA4 GPR132 LOC102723354 AHNAK2 JAG2 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | C14orf79 CDCA4 PLD4 AHNAK2 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | C14orf79 PLD4 AHNAK2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | LOC105370691 SNORA28 LINC00637 BRF1 PACS2 GPR132 SIVA1 BAG5 LOC101929634 RD3L EIF5 LOC105378183 ZFYVE21 INF2 LINC00638 TMEM179 ASPG MIR4710 C14orf79 C14orf2 CDCA4 TDRD9 MIR203A NUDT14 C14orf180 LOC388022 TRMT61A KIF26A PPP1R13B ZBTB42 LOC105370708 MIR203B AKT1 ADSSL1 BTBD6 CEP170B PLD4 LOC105370687 MIR6765 AHNAK2 MARK3 JAG2 KLC1 LOC107984648 XRCC3 CKB APOPT1 LOC102723354 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | C14orf79 BRF1 CDCA4 GPR132 BTBD6 NUDT14 LOC102723354 MIR6765 AHNAK2 JAG2 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | C14orf79 BRF1 CDCA4 GPR132 NUDT14 LOC102723354 MIR6765 AHNAK2 JAG2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |