Assay Details
Target Gene Details
Entrez Gene ID: | 91828 |
Gene Name: | exocyst complex component 3 like 4 |
Gene Aliases: |
C14orf73 |
Location: |
Chr.14:103094202-103110562 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L4 | NM_001077594.1 | NP_001071062.1 | ||
XM_011537323.2 | XP_011535625.1 | |||
XM_011537324.2 | XP_011535626.1 | |||
XM_011537325.2 | XP_011535627.1 | |||
XM_011537327.2 | XP_011535629.1 | |||
XM_011537328.2 | XP_011535630.1 | |||
XM_011537329.2 | XP_011535631.1 | |||
XM_011537330.2 | XP_011535632.1 | |||
XM_011537331.1 | XP_011535633.1 | |||
XM_011537332.2 | XP_011535634.1 | |||
XM_011537333.2 | XP_011535635.1 | |||
XM_011537334.1 | XP_011535636.1 | |||
AK000671.1 | ||||
BC113655.1 | ||||
BC117375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv565971 | Chr.14:103100567 - 103135315 on Build GRCh38 | Loss | TNFAIP2 EXOC3L4 LINC00677 |
nsv565970 | Chr.14:103089160 - 103139470 on Build GRCh38 | Loss | TNFAIP2 EXOC3L4 LINC00677 LOC107984640 |
nsv9168 | Chr.14:103065392 - 103446969 on Build GRCh38 | Loss | LINC00605 TNFAIP2 SNORA28 EXOC3L4 LINC00677 LOC105378183 MARK3 LOC107984640 LOC105370687 EIF5 |
nsv832882 | Chr.14:102970609 - 103142844 on Build GRCh38 | Loss | TNFAIP2 EXOC3L4 LINC00677 CDC42BPB LOC107984640 |
nsv952257 | Chr.14:103067564 - 103135963 on Build GRCh38 | Deletion | TNFAIP2 EXOC3L4 LINC00677 LOC107984640 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |