Assay Details
Target Gene Details
Entrez Gene ID: | 4140 |
Gene Name: | microtubule affinity regulating kinase 3 |
Gene Aliases: |
CTAK1, KP78, PAR1A, Par-1a |
Location: |
Chr.14:103385364-103503831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952258 | Chr.14:103386664 - 103405163 on Build GRCh38 | Duplication | MARK3 |
nsv9168 | Chr.14:103065392 - 103446969 on Build GRCh38 | Loss | TNFAIP2 SNORA28 LOC107984640 LINC00605 LINC00677 LOC105370687 MARK3 LOC105378183 EXOC3L4 EIF5 |
esv3635592 | Chr.14:103373908 - 103388946 on Build GRCh38 | Gain | MARK3 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | MIR4710 ZBTB42 LINC00638 TMEM179 LOC105370687 LOC105378183 INF2 CEP170B SIVA1 SNORA28 MIR6765 ASPG LOC105370708 LOC102723354 CDCA4 KIF26A ZFYVE21 JAG2 RD3L LOC388022 TRMT61A AKT1 CKB PPP1R13B XRCC3 ADSSL1 TDRD9 AHNAK2 LINC00637 NUDT14 C14orf2 BTBD6 PACS2 BRF1 LOC101929634 PLD4 BAG5 C14orf79 APOPT1 C14orf180 MARK3 LOC105370691 MIR203A GPR132 LOC107984648 MIR203B KLC1 EIF5 |
dgv422e214 | Chr.14:103355152 - 103437655 on Build GRCh38 | Gain | MARK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |