Assay Details
Target Gene Details
Entrez Gene ID: | 253970 |
Gene Name: | surfactant associated 3 |
Gene Aliases: |
NANCI, SFTPH, SP-H |
Location: |
Chr.14:36473288-36513785 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SFTA3 | NM_001101341.1 | NP_001094811.1 | ||
AY102071.1 | ||||
KF856952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832771 | Chr.14:36301801 - 36494836 on Build GRCh38 | Loss | MBIP DPPA3P2 SFTA3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |