Assay Details
Target Gene Details
Entrez Gene ID: | 85457 |
Gene Name: | CLOCK interacting pacemaker |
Gene Aliases: |
KIAA1737 |
Location: |
Chr.14:77098235-77117287 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CIPC | NM_033426.2 | NP_219494.2 | ||
AK096603.1 | ||||
AK315124.1 | ||||
AL834514.1 | CAD39170.1 | |||
BC037300.2 | AAH37300.1 | |||
BI546511.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832834 | Chr.14:77088644 - 77258014 on Build GRCh38 | Gain+Loss | CIPC ZDHHC22 TMEM63C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |