Assay Details
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | NP_001093588.1 | ||
NM_001100119.1 | NP_001093589.1 | |||
NM_005432.3 | NP_005423.1 | |||
XM_005268046.2 | XP_005268103.1 | |||
XM_011537138.2 | XP_011535440.1 | |||
AF035586.1 | AAC05368.1 | |||
AK022829.1 | ||||
AK023646.1 | ||||
AK124498.1 | ||||
AK126706.1 | 1 | 945 | ||
BC001036.1 | AAH01036.1 | |||
BC011725.2 | AAH11725.1 | |||
BT007417.1 | AAP36085.1 | |||
BX161398.1 | CAD61884.1 | |||
DB041321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | LOC107984648 XRCC3 KLC1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | ZFYVE21 LOC107984648 XRCC3 KLC1 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | ZFYVE21 LOC107984648 XRCC3 KLC1 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | ZFYVE21 LOC107984648 XRCC3 KLC1 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | C14orf180 MIR203B LOC107984648 GPR132 SNORA28 MIR203A PACS2 CDCA4 PLD4 BAG5 ASPG PPP1R13B EIF5 TRMT61A CEP170B ZFYVE21 JAG2 LOC102723354 BTBD6 TMEM179 KIF26A LOC105370708 LOC105370691 CKB ZBTB42 AKT1 LINC00637 BRF1 LINC00638 RD3L XRCC3 NUDT14 C14orf79 TDRD9 INF2 MIR4710 AHNAK2 C14orf2 LOC388022 LOC105370687 ADSSL1 KLC1 MARK3 LOC105378183 MIR6765 SIVA1 LOC101929634 APOPT1 |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | LINC00637 ZFYVE21 LOC107984648 RD3L XRCC3 KLC1 TDRD9 C14orf2 PPP1R13B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |