Assay Details
Target Gene Details
Entrez Gene ID: | 10858 |
Gene Name: | cytochrome P450 family 46 subfamily A member 1 |
Gene Aliases: |
CP46, CYP46 |
Location: |
Chr.14:99684262-99727321 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP46A1 | NM_006668.1 | NP_006659.1 | ||
XM_011536364.1 | XP_011534666.1 | |||
XM_017020933.1 | XP_016876422.1 | |||
AB209749.1 | BAD92986.1 | |||
AF094480.1 | AAD41244.1 | |||
AK295216.1 | ||||
BC022539.1 | AAH22539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv565733 | Chr.14:99522709 - 100097807 on Build GRCh38 | Gain | LOC102724682 EML1 EVL CYP46A1 HHIPL1 CCDC85C |
esv3635488 | Chr.14:99660725 - 99718739 on Build GRCh38 | Gain | CYP46A1 HHIPL1 |
nsv565763 | Chr.14:99683165 - 99767016 on Build GRCh38 | Loss | EML1 CYP46A1 |
nsv470667 | Chr.14:99658311 - 99742653 on Build GRCh38 | Loss | EML1 CYP46A1 HHIPL1 |
nsv1046296 | Chr.14:99640025 - 99723163 on Build GRCh38 | Gain | CYP46A1 HHIPL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |