Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_001318380.1 | NP_001305309.1 | ||
NM_177533.4 | NP_803877.2 | |||
AB087802.1 | BAC65455.1 | |||
AK128196.1 | ||||
AK310449.1 | ||||
BC041584.1 | AAH41584.1 | |||
DA249638.1 | ||||
DA409636.1 | ||||
DB096906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | MIR6765 JAG2 NUDT14 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | BRF1 TEX22 LOC102723354 CRIP2 PACS2 GPR132 LOC100507437 MTA1 MIR6765 BTBD6 JAG2 NUDT14 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | TEX22 CRIP1 PACS2 LOC100507437 MIR6765 C14orf80 NUDT14 LOC105370697 BRF1 CRIP2 IGH MTA1 IGHA2 BTBD6 JAG2 TMEM121 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | BRF1 LOC102723354 GPR132 MIR6765 JAG2 NUDT14 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | BRF1 MIR6765 JAG2 NUDT14 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | MIR6765 JAG2 NUDT14 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | C14orf2 RD3L PLD4 MARK3 MIR203A PACS2 INF2 PPP1R13B C14orf180 KIF26A LOC107984648 AKT1 MIR4710 LOC101929634 LINC00638 TRMT61A ASPG EIF5 APOPT1 BAG5 TMEM179 CEP170B TDRD9 CDCA4 LINC00637 C14orf79 AHNAK2 ZBTB42 MIR6765 LOC105378183 NUDT14 SNORA28 BRF1 LOC102723354 ZFYVE21 LOC105370687 GPR132 CKB LOC388022 LOC105370691 SIVA1 MIR203B KLC1 BTBD6 XRCC3 JAG2 LOC105370708 ADSSL1 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | BRF1 LOC102723354 CDCA4 C14orf79 GPR132 AHNAK2 MIR6765 BTBD6 JAG2 NUDT14 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | TEX22 ADAM6 LOC107987211 CRIP1 MIR4537 PACS2 IGHM LOC100507437 IGHV3-23 LINC00221 KIAA0125 IGHD MIR4507 IGHV3-41 IGHV3-69-1 IGHV3-71 IGHV3-30 TMEM121 MIR4539 IGHV4-31 IGHV1-69 IGHV4-55 LINC00226 MIR4538 IGHV3-48 IGHG1 MIR8071-2 MIR6765 C14orf80 IGHA1 IGHG2 NUDT14 LOC105370697 BRF1 IGHG3 IGHG4 CRIP2 IGH ELK2AP MTA1 MIR8071-1 IGHA2 BTBD6 JAG2 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | BRF1 LOC102723354 CDCA4 C14orf79 GPR132 AHNAK2 MIR6765 JAG2 NUDT14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |