Assay Details
Target Gene Details
Entrez Gene ID: | 51466 |
Gene Name: | Enah/Vasp-like |
Gene Aliases: |
RNB6 |
Location: |
Chr.14:99971454-100144236 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EVL | XM_017021364.1 | XP_016876853.1 | ||
AF112209.1 | AAF17197.1 | |||
AK289720.1 | ||||
BC023997.2 | AAH23997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv565733 | Chr.14:99522709 - 100097807 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv832873 | Chr.14:99820652 - 99997237 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)