Assay Details
Target Gene Details
Entrez Gene ID: | 3320 |
Gene Name: | heat shock protein 90 alpha family class A member 1 |
Gene Aliases: |
EL52, HEL-S-65p, HSP86, HSP89A, HSP90A, HSP90N, HSPC1, HSPCA, HSPCAL1, HSPCAL4, HSPN, Hsp89, Hsp90, LAP-2, LAP2 |
Location: |
Chr.14:102080738-102139749 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSP90AA1 | NM_001017963.2 | NP_001017963.2 | ||
XM_011536718.2 | XP_011535020.1 | |||
AJ890082.1 | CAI64495.1 | |||
AK056446.1 | ||||
BX248761.1 | CAD66568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv565947 | Chr.14:102001510 - 102204240 on Build GRCh38 | Gain | HSP90AA1 WDR20 DYNC1H1 |
nsv1428 | Chr.14:102068860 - 102102356 on Build GRCh38 | Insertion | HSP90AA1 |
dgv25n68 | Chr.14:102000993 - 102195522 on Build GRCh38 | Loss | HSP90AA1 WDR20 DYNC1H1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |