Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 | ||
XM_005267299.1 | XP_005267356.1 | |||
AK094143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | CDCA4 C14orf79 AHNAK2 GPR132 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | CDCA4 C14orf79 JAG2 AHNAK2 GPR132 LOC102723354 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | CDCA4 C14orf79 AHNAK2 PLD4 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | C14orf79 AHNAK2 PLD4 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | PPP1R13B LOC105370708 C14orf180 ADSSL1 PLD4 SIVA1 LOC388022 ASPG LOC105370691 MIR203A LOC105378183 PACS2 ZBTB42 KLC1 JAG2 INF2 LOC102723354 BAG5 BRF1 EIF5 KIF26A LINC00638 C14orf2 AHNAK2 CKB TRMT61A XRCC3 TMEM179 TDRD9 APOPT1 LOC101929634 ZFYVE21 CDCA4 MARK3 MIR203B SNORA28 LINC00637 C14orf79 LOC107984648 GPR132 MIR4710 BTBD6 NUDT14 LOC105370687 MIR6765 RD3L CEP170B AKT1 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | CDCA4 C14orf79 JAG2 AHNAK2 GPR132 BTBD6 NUDT14 LOC102723354 MIR6765 BRF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |