Assay Details
Target Gene Details
Entrez Gene ID: | 9369 |
Gene Name: | neurexin 3 |
Gene Aliases: |
C14orf60 |
Location: |
Chr.14:77979904-79868290 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NRXN3 | XM_017021790.1 | XP_016877279.1 | ||
XM_017021791.1 | XP_016877280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34345 | Chr.14:77904112 - 78030244 on Build GRCh38 | Loss | NRXN3 ADCK1 |
esv2759999 | Chr.14:77912813 - 78035676 on Build GRCh38 | Loss | NRXN3 ADCK1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |