Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_001318380.1 | NP_001305309.1 | ||
NM_177533.4 | NP_803877.2 | |||
AB087802.1 | BAC65455.1 | |||
AK128196.1 | ||||
AK310449.1 | ||||
BC041584.1 | AAH41584.1 | |||
DA249638.1 | ||||
DA409636.1 | ||||
DB096906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | JAG2 NUDT14 MIR6765 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | MTA1 BTBD6 CRIP2 LOC102723354 JAG2 NUDT14 GPR132 PACS2 TEX22 LOC100507437 BRF1 MIR6765 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | IGHA2 BTBD6 CRIP2 JAG2 PACS2 CRIP1 TEX22 C14orf80 LOC105370697 MTA1 NUDT14 IGH TMEM121 LOC100507437 BRF1 MIR6765 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | LOC102723354 JAG2 NUDT14 GPR132 BRF1 MIR6765 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | JAG2 NUDT14 BRF1 MIR6765 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | JAG2 NUDT14 MIR6765 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | EIF5 LOC107984648 AKT1 PACS2 KLC1 TRMT61A ZBTB42 TDRD9 CDCA4 BAG5 PLD4 LINC00637 LOC101929634 LOC105370708 MIR203B C14orf2 CEP170B BRF1 LOC388022 LINC00638 BTBD6 PPP1R13B LOC105370691 MIR203A SIVA1 JAG2 INF2 KIF26A ADSSL1 MARK3 LOC105378183 AHNAK2 MIR4710 SNORA28 TMEM179 LOC105370687 XRCC3 C14orf180 LOC102723354 RD3L CKB ASPG NUDT14 GPR132 APOPT1 C14orf79 ZFYVE21 MIR6765 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | CDCA4 BTBD6 LOC102723354 JAG2 NUDT14 GPR132 C14orf79 AHNAK2 BRF1 MIR6765 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | IGHG1 IGHA2 IGHG2 CRIP2 IGHV1-69 PACS2 MIR4507 IGHD KIAA0125 MTA1 IGHV3-23 LINC00221 LOC107987211 MIR4537 ELK2AP IGHV3-41 IGHA1 BRF1 IGHG4 IGHV3-69-1 BTBD6 JAG2 ADAM6 LINC00226 MIR4538 CRIP1 TEX22 IGHM IGHV3-71 C14orf80 LOC105370697 IGHV3-30 MIR8071-2 MIR8071-1 IGHV4-31 IGHG3 MIR4539 NUDT14 IGH TMEM121 IGHV3-48 LOC100507437 IGHV4-55 MIR6765 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | CDCA4 LOC102723354 JAG2 NUDT14 GPR132 C14orf79 AHNAK2 BRF1 MIR6765 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |