Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 | ||
XM_005267299.1 | XP_005267356.1 | |||
AK094143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | AHNAK2 C14orf79 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | AHNAK2 CDCA4 GPR132 C14orf79 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | LOC102723354 AHNAK2 CDCA4 GPR132 C14orf79 JAG2 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | PLD4 AHNAK2 CDCA4 C14orf79 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | PLD4 AHNAK2 C14orf79 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | APOPT1 LINC00638 LOC105370708 ASPG CDCA4 LOC388022 KLC1 LOC105370691 LOC107984648 ZBTB42 PLD4 CEP170B PACS2 KIF26A TRMT61A SNORA28 GPR132 AKT1 C14orf79 C14orf180 ZFYVE21 LOC105378183 TDRD9 LOC101929634 PPP1R13B LINC00637 MIR4710 CKB MIR203A INF2 SIVA1 BAG5 XRCC3 MIR203B BTBD6 NUDT14 C14orf2 BRF1 LOC102723354 LOC105370687 RD3L ADSSL1 AHNAK2 MARK3 MIR6765 TMEM179 EIF5 JAG2 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | LOC102723354 AHNAK2 MIR6765 CDCA4 GPR132 BTBD6 NUDT14 C14orf79 BRF1 JAG2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |