Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_177533.4 | NP_803877.2 | ||
AB087802.1 | BAC65455.1 | |||
AK128196.1 | 6 | 1489 | ||
BC041584.1 | AAH41584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | MIR6765 JAG2 NUDT14 |
nsv566177 | Chr.14:105167384 - 105177639 on Build GRCh38 | Gain | JAG2 NUDT14 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | GPR132 CRIP2 MIR6765 TEX22 JAG2 BTBD6 MTA1 LOC100507437 NUDT14 LOC102723354 BRF1 PACS2 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | IGH LOC105370697 IGHA2 PACS2 CRIP2 CRIP1 MIR6765 TEX22 JAG2 C14orf80 BTBD6 MTA1 LOC100507437 TMEM121 NUDT14 BRF1 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | GPR132 MIR6765 JAG2 NUDT14 LOC102723354 BRF1 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | MIR6765 JAG2 NUDT14 BRF1 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | MIR6765 JAG2 NUDT14 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | AKT1 XRCC3 PPP1R13B TRMT61A SIVA1 MIR4710 ZBTB42 SNORA28 LOC105370708 LINC00637 LOC388022 ZFYVE21 CKB LINC00638 GPR132 C14orf79 AHNAK2 INF2 LOC101929634 BAG5 MIR203A JAG2 CEP170B APOPT1 C14orf2 NUDT14 LOC102723354 ASPG KIF26A BRF1 RD3L EIF5 ADSSL1 LOC105370687 MIR203B C14orf180 PACS2 MARK3 TDRD9 LOC107984648 KLC1 MIR6765 CDCA4 BTBD6 LOC105370691 PLD4 LOC105378183 TMEM179 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | GPR132 C14orf79 AHNAK2 MIR6765 CDCA4 JAG2 BTBD6 NUDT14 LOC102723354 BRF1 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | IGH LOC105370697 ELK2AP IGHV3-71 IGHV3-48 MIR8071-2 TEX22 IGHD LINC00226 IGHV4-31 JAG2 C14orf80 LOC100507437 IGHV3-30 TMEM121 NUDT14 BRF1 IGHM IGHA2 IGHA1 IGHV3-69-1 ADAM6 LOC107987211 PACS2 IGHG1 MIR4539 CRIP2 CRIP1 MIR6765 IGHG4 IGHG3 MIR4507 IGHV3-23 KIAA0125 IGHV4-55 BTBD6 MTA1 LINC00221 IGHV1-69 IGHG2 MIR8071-1 MIR4537 MIR4538 IGHV3-41 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | GPR132 C14orf79 AHNAK2 MIR6765 CDCA4 JAG2 NUDT14 LOC102723354 BRF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |