Assay Details
Target Gene Details
Entrez Gene ID: | 196913 |
Gene Name: | long intergenic non-protein coding RNA 1599 |
Gene Aliases: |
C14orf183 |
Location: |
Chr.14:50007313-50105043 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01599 | NR_131171.1 | |||
BF691660.1 | ||||
BQ027525.1 | ||||
BU927649.1 | ||||
T90102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss | LINC01588 RN7SL1 MAP4K5 KLHDC2 CDKL1 LOC100506446 LOC100288910 SOS2 MIR4504 POLE2 MIR6076 LRR1 L2HGDH RPL36AL KLHDC1 RPS29 DNAAF2 VCPKMT ATP5S LINC01599 RN7SL2 MGAT2 ARF6 NEMF |