Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | AHNAK2 C14orf79 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | AHNAK2 CDCA4 C14orf79 GPR132 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | JAG2 AHNAK2 CDCA4 C14orf79 GPR132 LOC102723354 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | PLD4 AHNAK2 CDCA4 C14orf79 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | PLD4 AHNAK2 C14orf79 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | EIF5 RD3L GPR132 XRCC3 ZFYVE21 LOC107984648 ZBTB42 LOC105370708 LOC101929634 PLD4 TMEM179 CDCA4 INF2 PACS2 NUDT14 LOC102723354 KLC1 CEP170B C14orf180 ASPG AKT1 MIR203B LINC00638 AHNAK2 LOC105378183 C14orf2 MARK3 SNORA28 PPP1R13B CKB TRMT61A MIR203A JAG2 APOPT1 SIVA1 BTBD6 ADSSL1 TDRD9 C14orf79 LOC105370687 KIF26A LOC105370691 MIR6765 LOC388022 BAG5 MIR4710 LINC00637 BRF1 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | JAG2 AHNAK2 BTBD6 CDCA4 C14orf79 NUDT14 GPR132 LOC102723354 MIR6765 BRF1 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | JAG2 AHNAK2 CDCA4 C14orf79 NUDT14 GPR132 LOC102723354 MIR6765 BRF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |