Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 | ||
XM_005267299.1 | XP_005267356.1 | |||
AK094143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | C14orf79 CDCA4 GPR132 AHNAK2 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | C14orf79 PLD4 CDCA4 AHNAK2 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | C14orf79 PLD4 AHNAK2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | TRMT61A EIF5 APOPT1 LOC105370708 TDRD9 PACS2 XRCC3 C14orf180 ZFYVE21 LOC102723354 ADSSL1 MIR6765 INF2 AKT1 C14orf2 LOC107984648 RD3L LOC101929634 MARK3 AHNAK2 ASPG LINC00637 ZBTB42 TMEM179 NUDT14 CEP170B LOC105370687 PLD4 BRF1 PPP1R13B SIVA1 LOC105370691 MIR203B LOC388022 C14orf79 KIF26A BAG5 CKB MIR203A CDCA4 MIR4710 LINC00638 GPR132 SNORA28 LOC105378183 JAG2 BTBD6 KLC1 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | C14orf79 NUDT14 CDCA4 BRF1 GPR132 AHNAK2 LOC102723354 JAG2 MIR6765 BTBD6 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |