Assay Details
Target Gene Details
Entrez Gene ID: | 55504 |
Gene Name: | TNF receptor superfamily member 19 |
Gene Aliases: |
TAJ, TAJ-alpha, TRADE, TROY |
Location: |
Chr.13:23570248-23676105 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNFRSF19 | NM_001204458.1 | NP_001191387.1 | ||
NM_001204459.1 | NP_001191388.1 | |||
NM_018647.3 | NP_061117.2 | |||
NM_148957.3 | NP_683760.1 | |||
XM_005266445.2 | XP_005266502.1 | |||
XM_005266446.1 | XP_005266503.1 | |||
XM_011535146.1 | XP_011533448.1 | |||
XM_017020651.1 | XP_016876140.1 | |||
XM_017020652.1 | XP_016876141.1 | |||
AF167555.1 | AAF71828.1 | |||
AK292874.1 | ||||
AK304290.1 | ||||
AY358888.1 | AAQ89247.1 | |||
BC047321.1 | AAH47321.1 | |||
DA192370.1 | ||||
DB223760.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525965 | Chr.13:22937442 - 24358507 on Build GRCh38 | Gain | SPATA13 LINC00327 TNFRSF19 SACS C1QTNF9B SPATA13-AS1 NUS1P3 LINC00566 MIR2276 MIPEP SACS-AS1 ANKRD20A19P C1QTNF9 SGCG C1QTNF9B-AS1 |
esv3631523 | Chr.13:23479493 - 23611329 on Build GRCh38 | Gain | LINC00327 TNFRSF19 |
nsv561292 | Chr.13:23025847 - 24348376 on Build GRCh38 | Loss | SPATA13 LINC00327 TNFRSF19 SACS C1QTNF9B SPATA13-AS1 NUS1P3 LINC00566 MIR2276 MIPEP SACS-AS1 ANKRD20A19P C1QTNF9 SGCG C1QTNF9B-AS1 |
dgv3084n54 | Chr.13:22925582 - 24378278 on Build GRCh38 | Gain | SPATA13 LINC00327 TNFRSF19 SACS C1QTNF9B SPATA13-AS1 NUS1P3 LINC00566 MIR2276 MIPEP SACS-AS1 ANKRD20A19P C1QTNF9 SGCG C1QTNF9B-AS1 |
esv3892319 | Chr.13:23325488 - 23598910 on Build GRCh38 | Gain | LINC00327 SACS-AS1 TNFRSF19 SACS |
esv3892311 | Chr.13:22674660 - 24411768 on Build GRCh38 | Gain | SPATA13 LINC00327 TNFRSF19 LOC105370109 SACS C1QTNF9B SPATA13-AS1 NUS1P3 LINC00566 MIR2276 MIPEP SACS-AS1 ANKRD20A19P C1QTNF9 SGCG BASP1P1 C1QTNF9B-AS1 LINC00621 |
nsv832565 | Chr.13:23462415 - 23640719 on Build GRCh38 | Gain | LINC00327 TNFRSF19 |
esv3892320 | Chr.13:23486150 - 23650003 on Build GRCh38 | Loss | LINC00327 TNFRSF19 |
dgv1615n100 | Chr.13:22930769 - 24362849 on Build GRCh38 | Gain | SPATA13 LINC00327 TNFRSF19 SACS C1QTNF9B SPATA13-AS1 NUS1P3 LINC00566 MIR2276 MIPEP SACS-AS1 ANKRD20A19P C1QTNF9 SGCG C1QTNF9B-AS1 |
esv2751136 | Chr.13:22982261 - 24393377 on Build GRCh38 | Loss | SPATA13 LINC00327 TNFRSF19 SACS C1QTNF9B SPATA13-AS1 NUS1P3 LINC00566 MIR2276 MIPEP SACS-AS1 ANKRD20A19P C1QTNF9 SGCG C1QTNF9B-AS1 |
esv3580576 | Chr.13:22945778 - 24362710 on Build GRCh38 | Loss | SPATA13 LINC00327 TNFRSF19 SACS C1QTNF9B SPATA13-AS1 NUS1P3 LINC00566 MIR2276 MIPEP SACS-AS1 ANKRD20A19P C1QTNF9 SGCG C1QTNF9B-AS1 |
esv2422502 | Chr.13:22755560 - 24382791 on Build GRCh38 | Duplication | SPATA13 LINC00327 TNFRSF19 LOC105370109 SACS C1QTNF9B SPATA13-AS1 NUS1P3 LINC00566 MIR2276 MIPEP SACS-AS1 ANKRD20A19P C1QTNF9 SGCG BASP1P1 C1QTNF9B-AS1 LINC00621 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |