Assay Details
Target Gene Details
Entrez Gene ID: | 730249 |
Gene Name: | aconitate decarboxylase 1 |
Gene Aliases: |
CAD, IRG1 |
Location: |
Chr.13:76948559-76958642 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACOD1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832653 | Chr.13:76948899 - 77115221 on Build GRCh38 | Loss | CLN5 ACOD1 FBXL3 MYCBP2 MYCBP2-AS1 |
nsv832651 | Chr.13:76788718 - 76974704 on Build GRCh38 | Gain | BTF3P11 KCTD12 ACOD1 |
nsv832652 | Chr.13:76855107 - 77041507 on Build GRCh38 | Gain | BTF3P11 CLN5 KCTD12 ACOD1 FBXL3 |