Assay Details
Target Gene Details
Entrez Gene ID: | 2621 |
Gene Name: | growth arrest specific 6 |
Gene Aliases: |
AXLLG, AXSF |
Location: |
Chr.13:113820549-113864103 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAS6 | NM_000820.3 | NP_000811.1 | ||
AK122969.1 | ||||
AK126533.1 | BAC86580.1 | |||
AK290803.1 | ||||
AK303145.1 | ||||
BC038984.1 | AAH38984.1 | |||
L13720.1 | AAA58494.1 |
Target Gene Details
Entrez Gene ID: | 650669 |
Gene Name: | GAS6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:113815610-113842841 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAS6-AS1 | NR_044995.2 | |||
DA460725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv563390 | Chr.13:113809317 - 113845944 on Build GRCh38 | Loss | GAS6 GAS6-AS1 TMEM255B |
nsv1043348 | Chr.13:113756321 - 113867896 on Build GRCh38 | Gain | LOC105377805 GAS6-AS2 GAS6 GAS6-AS1 TMEM255B |
nsv951933 | Chr.13:113808028 - 113844427 on Build GRCh38 | Deletion | GAS6 GAS6-AS1 TMEM255B |
dgv3450n54 | Chr.13:113754200 - 113886735 on Build GRCh38 | Gain | LOC105377805 GAS6-AS2 LINC00454 GAS6 GAS6-AS1 TMEM255B |
esv24756 | Chr.13:113802553 - 113834906 on Build GRCh38 | Gain+Loss | GAS6 GAS6-AS1 TMEM255B |
nsv832727 | Chr.13:113753115 - 113918257 on Build GRCh38 | Loss | LOC105377805 GAS6-AS2 LINC00454 GAS6 GAS6-AS1 TMEM255B LINC00452 |
nsv563387 | Chr.13:113832338 - 113876058 on Build GRCh38 | Loss | GAS6-AS2 GAS6 GAS6-AS1 |
esv3582860 | Chr.13:113756381 - 113867871 on Build GRCh38 | Gain | LOC105377805 GAS6-AS2 GAS6 GAS6-AS1 TMEM255B |
nsv826836 | Chr.13:113753163 - 113875076 on Build GRCh38 | Loss | LOC105377805 GAS6-AS2 GAS6 GAS6-AS1 TMEM255B |
nsv826837 | Chr.13:113802020 - 113834906 on Build GRCh38 | Loss | GAS6 GAS6-AS1 TMEM255B |
esv3633622 | Chr.13:113808937 - 113835264 on Build GRCh38 | Gain | GAS6 GAS6-AS1 TMEM255B |
esv3633615 | Chr.13:113724844 - 113843481 on Build GRCh38 | Gain | LOC105377805 GAS6 GAS6-AS1 TMEM255B LINC00552 GRK1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |