Assay Details
Target Gene Details
Entrez Gene ID: | 101929140 |
Gene Name: | uncharacterized LOC101929140 |
Gene Aliases: |
- |
Location: |
Chr.13:41132928-41236686 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929140 | NR_120423.1 | |||
AK056182.1 | ||||
DA634395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832587 | Chr.13:41030795 - 41224672 on Build GRCh38 | Gain | MIR3168 WBP4 ELF1 LOC101929140 KBTBD7 MTRF1 KBTBD6 |
esv3631867 | Chr.13:41144449 - 41179962 on Build GRCh38 | Gain | LOC101929140 |
esv2747299 | Chr.13:41131549 - 41193448 on Build GRCh38 | Deletion | LOC101929140 KBTBD7 KBTBD6 |
nsv523358 | Chr.13:41144691 - 41186811 on Build GRCh38 | Loss | LOC101929140 |
nsv528180 | Chr.13:41141146 - 41186811 on Build GRCh38 | Gain | LOC101929140 |