Assay Details
Target Gene Details
Entrez Gene ID: | 647174 |
Gene Name: | serpin family E member 3 |
Gene Aliases: |
- |
Location: |
Chr.13:51334976-51362391 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINE3 | NM_001101320.1 | NP_001094790.1 | ||
XM_017020707.1 | XP_016876196.1 | |||
AM402969.1 | ||||
AX772926.1 | CAE06272.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832611 | Chr.13:51192336 - 51364178 on Build GRCh38 | Loss | FAM124A MIR5693 SERPINE3 INTS6 |
nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss | SPRYD7 DLEU2 MIR3613 FAM124A DLEU1-AS1 TRIM13 DLEU7 MIR5693 SERPINE3 DLEU1 MIR15A RNASEH2B-AS1 KCNRG INTS6-AS1 MIR16-1 DLEU7-AS1 GUCY1B2 ST13P4 INTS6 RNASEH2B LINC00371 |
nsv1041 | Chr.13:51325852 - 51352515 on Build GRCh38 | Insertion | MIR5693 SERPINE3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |