Assay Details
Target Gene Details
Entrez Gene ID: | 348013 |
Gene Name: | transmembrane protein 255B |
Gene Aliases: |
FAM70B |
Location: |
Chr.13:113759217-113813797 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM255B | NM_182614.2 | NP_872420.1 | ||
XM_005272413.3 | XP_005272470.1 | |||
XM_005272415.2 | XP_005272472.1 | |||
XM_006719986.2 | XP_006720049.1 | |||
XM_017020558.1 | XP_016876047.1 | |||
XM_017020559.1 | XP_016876048.1 | |||
XM_017020560.1 | XP_016876049.1 | |||
BC018995.2 | AAH18995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951932 | Chr.13:113782828 - 113807827 on Build GRCh38 | Deletion |
![]() |
nsv1043348 | Chr.13:113756321 - 113867896 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
dgv3450n54 | Chr.13:113754200 - 113886735 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv832727 | Chr.13:113753115 - 113918257 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3582860 | Chr.13:113756381 - 113867871 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv826836 | Chr.13:113753163 - 113875076 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3633615 | Chr.13:113724844 - 113843481 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)