Assay Details
Target Gene Details
Entrez Gene ID: | 348013 |
Gene Name: | transmembrane protein 255B |
Gene Aliases: |
FAM70B |
Location: |
Chr.13:113759217-113813797 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM255B | NM_182614.2 | NP_872420.1 | ||
XM_005272413.3 | XP_005272470.1 | |||
XM_005272415.2 | XP_005272472.1 | |||
XM_006719986.2 | XP_006720049.1 | |||
XM_017020558.1 | XP_016876047.1 | |||
XM_017020559.1 | XP_016876048.1 | |||
XM_017020560.1 | XP_016876049.1 | |||
BC018995.2 | AAH18995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1043348 | Chr.13:113756321 - 113867896 on Build GRCh38 | Gain | TMEM255B GAS6-AS1 LOC105377805 GAS6-AS2 GAS6 |
dgv3450n54 | Chr.13:113754200 - 113886735 on Build GRCh38 | Gain | TMEM255B GAS6-AS1 LINC00454 LOC105377805 GAS6-AS2 GAS6 |
nsv951931 | Chr.13:113774628 - 113780127 on Build GRCh38 | Deletion | TMEM255B |
nsv832727 | Chr.13:113753115 - 113918257 on Build GRCh38 | Loss | TMEM255B GAS6-AS1 LINC00454 LOC105377805 GAS6-AS2 GAS6 LINC00452 |
esv3582860 | Chr.13:113756381 - 113867871 on Build GRCh38 | Gain | TMEM255B GAS6-AS1 LOC105377805 GAS6-AS2 GAS6 |
nsv826836 | Chr.13:113753163 - 113875076 on Build GRCh38 | Loss | TMEM255B GAS6-AS1 LOC105377805 GAS6-AS2 GAS6 |
esv3633618 | Chr.13:113757581 - 113789485 on Build GRCh38 | Gain | TMEM255B LOC105377805 |
esv3633615 | Chr.13:113724844 - 113843481 on Build GRCh38 | Gain | TMEM255B GAS6-AS1 LOC105377805 LINC00552 GRK1 GAS6 |